Would you like to log in again?
Her2 Substrate,Biotin
Supplier: Enzo Life Sciences
BML-P243-0001
89161-142EA 299.72 USD InStock
89161-142
Her2 Substrate,Biotin
Proteins and Peptides
This peptide, Biotin-Ahx-GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty