Would you like to log in again?
Fgfr Substrate,Biotin
Supplier: Enzo Life Sciences
BML-P245-0001
89161-146EA 272.14 USD InStock
89161-146
Fgfr Substrate,Biotin
Proteins and Peptides
This peptide, Biotin-Ahx-KEAPEDLYKDFLTL, is derived from the KDR sequence and is a substrate for FGFR. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty