Would you like to log in again?
Peptide A
Supplier: Enzo Life Sciences
BML-P306-0500
89161-206EA 256.73 USD InStock
89161-206
Peptide A
Proteins and Peptides
Src kinase inhibitor
Sequence is pp60v-src (137-157), which is located in the SH3/SH2 domain region. Non-competitive inhibitor of Src and EGFR. Selectively inhibits protein tyrosine phosphorylation in cell extracts. Peptide is not phosphorylated by Src. IC50=7.5 µM for v-Src. Inhibitory target seems to be a catalytic negative regulatory domain of the tyrosine kinase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty