Would you like to log in again?
Insulin Receptor (1142-1153)
Supplier: Enzo Life Sciences
BML-P314-0500
89161-216EA 166.38 USD InStock
89161-216
Insulin Receptor (1142-1153)
Proteins and Peptides
Insulin receptor substrate
This sequence, TRDIYETDYYRK, is from the insulin receptor β-subunit cytoplasmic domain, inclusive of regulatory autophosphorylation sites Tyr1146, Tyr1150 and Tyr1151. It has been used as an insulin receptor tyrosine kinase substrate and the phosphorylated peptide as a protein tyrosine phosphatase substrate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty