Would you like to log in again?
[pTyr1146/1150] Insulin receptor (1142-1153)
Supplier: Enzo Life Sciences
BML-P321-0500
89161-230EA 210.46 USD InStock
89161-230
[pTyr1146/1150] Insulin receptor (1142-1153)
Proteins and Peptides
Insulin receptor substrate
This sequence, TRDIpYETDpYYRK, is from the insulin receptor ß-subunit cytoplasmic domain, inclusive of regulatory autophosphorylation sites Tyr1146, Tyr1150 and Tyr1151. The unphosphorylated version of this peptide has been used as an insulin receptor kinase substrate. The phosphorylated peptide has been used as a protein tyrosine phosphatase substrate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty