Would you like to log in again?
Salmon Calcitonin
Supplier: Enzo Life Sciences
BML-P420-0001 BML-P420-0005
89161-242EA 156.34 USD InStock
89161-242 89161-244
Salmon Calcitonin
Proteins and Peptides
Reduces blood calcium
Calcium-regulating peptide hormone released from the thyroid. Lowers calcium concentration in the blood and decreases bone resorption by osteoclasts. Used for therapeutic applications in osteoporosis. Salmon calcitonin peptide sequence differs from mammalian but is more potent.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty