Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Elastatinal ≥98%

Supplier: Enzo Life Sciences

BML-PI103-0005 BML-PI103-0025
89161-572EA 488 USD InStock
89161-572 89161-570
Elastatinal ≥98%
Elastatinal
Elastase inhibitor

Irreversible inhibitor of neutrophil and rat microglia elastase. Inhibits proteolytic activity of poliovirus and rhinovirus 2A proteinases.


Formula: C₂₁H₃₆N₈O₇
MW: 512.60 g/mol
MDL Number: MFCD00151180
CAS Number: 51798-45-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/39/45401439.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty