Would you like to log in again?
E-64-C
Supplier: Enzo Life Sciences
BML-PI106-0005 BML-PI106-0001
89161-580EA 386.2 USD InStock
89161-580 89161-578
E-64-C
Proteins and Peptides
Calpain and cathepsin inhibitor
Cell-impermeable epoxysuccinyl peptide inhibitor of calpain and other cysteine proteases. Inhibits calpain and cathepsins B, H, and L.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty