Would you like to log in again?
Proteasome ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
PW0575-0001
89161-908EA 614.83 USD InStock
89161-908
Proteasome ELISA Kit, Enzo Life Sciences
Assays ELISAs
This kit provides the means to quantify 20S proteasome concentrations in biological samples using a sandwich ELISA technique, utilizing two 20S proteasome specific antibodies for capture and detection purposes together with a highly sensitive substrate
  • Determination of 20S proteasome levels in biological samples (cell lysates, tissue extracts, plasma, serum)
  • Comparison of 20S proteasome levels in plasma/serum samples associated with a particular disease/illness with samples from healthy controls
  • Investigation of variation in 20S proteasome levels in abnormal cell lines/tissues
  • Assay Time: <3.5 hours
Sample 20S proteasome levels are determined by comparison to a 20S proteasome calibration curve produced in parallel. This kit provides sufficient material for 1x96 well plate set-up to be run.
Delivery information: Kit consists of 96 wells (24 tests in triplicate), plate, capture antibody, detection antibody, conjugated antibody, Proteasome stock solution, binding buffer, wash buffer, blocking buffer, ELISA buffer, lysis buffer, TMB substrate solution, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty