Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ubiquitin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW0600-0025 PW0600-0100
89161-924EA 495.81 USD InStock
89161-924 89161-922
Anti-Ubiquitin Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes K63-linked polyubiquitin in wide range of species. Does not cross-react with any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitinylated species.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty