Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMA4 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8120-0025 PW8120-0100
89161-998EA 526.7 USD InStock
89161-998 89161-996
Anti-PSMA4 Mouse Monoclonal Antibody
Antibodies
Two forms of the protein are produced by alternative splicing of the gene - subunits XAPC7-S and XAPC7-L.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human proteasome 20S (alpha4 subunit).

Type: Primary
Antigen: PSMA4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty