Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SQ-29548 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-RA103-0001
89162-472EA 120.36 USD InStock
89162-472
SQ-29548 ≥98% (by HPLC)
SQ-29548
Potent, selective thromboxane A₂ receptor antagonist. Shows inhibition of human platelet aggregation initiated by arachidonic acid (IC₅₀=12 nM) . Ki=7.9 nM for inhibition of [³H]-U-46619 binding to human platelet membranes.

Soluble in 100% ethanol, methanol or acetonitrile; also soluble in PBS, pH 7.2, DMSO or dimethyl formamide.

Synonym: [1S-[1α,2α(Z),3α,4α]]-7-[3-[[2-[(Phenylamino)carbonyl]hydrazino]methyl]-7-oxabicyclo[2.2.1]hept-2-yl]-5-heptanoic acid]


Formula: C₂₁H₂₉N₃O₄
MW: 387.5 g/mol
Storage Temperature: Freezer
CAS Number: 98672-91-4
Order Now
Specification Test Results
Purity ≥98% (HPLC)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty