Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nmda Receptor NR1 Subunit Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
BML-SA493-0015
89163-190EA 546.42 USD InStock
89163-190
Anti-Nmda Receptor NR1 Subunit Mouse Monoclonal Antibody
Antibodies
Host: Mouse

  • For Immunoprecipitation, Western Blot

Recognizes mouse and rat NMDA receptor (NR1 subunit).

Type: Primary
Antigen: NMDA RECEPTOR NR1 SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty