Would you like to log in again?
Human Recombinant MMP-13 (catalytic domain) (from E. coli)
Supplier: Enzo Life Sciences
BML-SE246-0010
89163-888EA 563.23 USD InStock
89163-888
Human Recombinant MMP-13 (catalytic domain) (from E. coli)
Proteins and Peptides
Produced in E. coli. Active Matrix Metalloproteinase-13 (MMP-13, collagenase-3) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-13 (Tyr104-Asn274, NM_002427) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-13 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty