Would you like to log in again?
Human Recombinant Cathepsin S
Supplier: Enzo Life Sciences
BML-SE453-0010
89164-320EA 540.76 USD InStock
89164-320
Human Recombinant Cathepsin S
Proteins and Peptides
Highly active
Produced in insect cells. Recombinant active cathepsin S cloned from human cDNA as full length proenzyme, auto-activated by incubation in pH 4.5 acetate buffer, and purified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty