Would you like to log in again?
Human Recombinant MMP-13
Supplier: Enzo Life Sciences
BML-SE493-0010
89164-388EA 538.51 USD InStock
89164-388
Human Recombinant MMP-13
Proteins and Peptides
Produced in insect cells. Primarily full length latent MMP-13, with some active forms present.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty