Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C8 ceramide ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-SL112-0005 BML-SL112-0025
89164-586EA 348 USD InStock
89164-586 89164-584
C8 ceramide ≥98% (by TLC)
C8 ceramide
Ceramide-activated protein kinase activator.

A biologically active, cell-permeable, but nonphysiologic ceramide analog. It induces phosphorylation of epidermal growth factor receptor on Thr-669 in A-431 cells by stimulation of ceramide-activated protein kinase and induces apoptosis.

Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).


Formula: C₂₆H₅₁NO₃
MW: 425.698 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236496
CAS Number: 74713-59-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/12/45401512.jpg
Order Now
Specification Test Results
Purity ≥98% (TLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty