Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palmityltrifluoromethylketone ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST336-0010 BML-ST336-0050
89164-810EA 94.75 USD InStock
89164-810 89164-812
Palmityltrifluoromethylketone ≥98% (by TLC)
Palmityltrifluoromethylketone
Inhibitor of both Ca2+-dependent cytosolic cPLA2 (IC50=45 µM) and Ca2+-independent phospholipases A2 iPLA2 (IC50=3.8 µM).

  • Phospholipase inhibitor

White to off-white solid.

Soluble in 100% ethanol, dimethyl formamide or DMSO (warm); sparingly soluble in neutral buffers.


Formula: C₁₇H₃₁F₃O
MW: 308.429 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00797669
CAS Number: 141022-99-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/36/45401536.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty