Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NGAL (Dog) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-043
89133-386EA 1075.13 USD InStock
89133-386
NGAL (Dog) ELISA Kit, Enzo Life Sciences
Assays ELISAs
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in dog samples
  • Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
For the quantitative determination of NGAL in dog urine, plasma, serum, tissue extracts, or culture supernatants.
Absorbance is read at 450 nm.
Delivery information: Kit consists of microwell plate coated with anti-NGAL (dog) antibody (96 wells), 5X sample diluent, NGAL Dog calibrators 1-8; 0, 4, 10, 20, 40, 100, 200, 400 pg/mL, 25X wash solution concentrate, biotinylated Dog NGAL antibody, HRP-Streptavidin, TMB substrate, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty