Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Macrophage Marker Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ENZ-30935
89165-680EA 294 USD InStock
89165-680
Anti-Macrophage Marker Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For Immunohistochemistry

Recognizes human and monkey fixed macrophages such as "tingible macrophages", interdigitating macrophages of lymph nodes or tissue macrophages. Cross-reacts with a subpopulation of endothelial cells including those of capillaries and smaller blood vessels. Does not cross-react with B or T cells.

Type: Primary
Antigen: MACROPHAGE MARKER
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty