Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indo-1 AM ≥95% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences
Synonyms: 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylesterIndo-1-(acetoxymethyl) esterIndo-1 AM Ester

ENZ-52008
89165-946EA 223 USD InStock
89165-946
Indo-1 AM ≥95% (by HPLC), Ultrapure
Indo-1 AM
Calcium dye

Indo-1 acetomethoxy (AM) is a popular UV-excitable calcium indicator, similar to Fura-2. In contrast to Fura-2, Indo-1 has a dual emission peak. The main emission peak in calcium-free solution is 475nm while in the presence of calcium the emission is shifted to 400nm. Indo-1 AM is a membrane permeable probe and can be easily loaded into cells by passive incubation. The probe is widely used in flow cytometry. For this application, it is critical to have an instrument with an argon laser tuned to UV or a heliumcadmium laser available. Wavelength Maxima: Excitation 346nm, Emission 475nm


Formula: C₄₇H₅₁N₃O₂₂
MW: 1009.91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036955
CAS Number: 112926-02-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/90/45401590.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty