Would you like to log in again?
JC-10 ≥95% (by HPLC), Ultrapure
Supplier: Enzo Life Sciences
Synonyms: Enhanced JC-1
ENZ-52305
89166-016EA 298 USD InStock
89166-016
JC-10 ≥95% (by HPLC), Ultrapure
JC-10
Mitochondria dye
JC-10 is a derivative of JC-1 useful for determining mitochondrial membrane potential by flow cytometry, fluorescence microscopy and in microplate-based fluorescent assays. JC-10 accumulates in mitochondria, selectively generating an orange J-aggregate emission profile (590 nm) in healthy cells. However, upon cell injury, as membrane potential decreases, JC-10 monomers are generated, resulting in a shift to green emission (525 nm). The principal advantages of JC-10 relative to JC-1 include improved solubility in aqueous media and an ability to detect subtler changes in mitochondrial membrane potential loss. JC-10 allows for both qualitative visualization, considering the shift from orange to green fluorescence emission, and quantitative detection, considering the fluorescence intensity ratio, of mitochondrial membrane potential changes. Wavelength Maxima: Excitation 510nm, Emission 525nm
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty