Would you like to log in again?
Solid State Voltage Controller, 0 – 120 V at 10 Amps, Ace Glass Incorporated
Supplier: Ace Glass
12087-10
89076-332EA 1364.67 USD InStock
89076-332
Solid State Voltage Controller, 0 – 120 V at 10 Amps, Ace Glass Incorporated
Controllers Utility Controllers
This Solid State Voltage Controller offers variable control from zero output to 95% line voltage.
A voltage-limit, rear-mounted center-off switch is used to select a 40 V or 120 V maximum output. With 0 – 10 ammeter, and 5% accuracy. Warning glow light mounted next to correcting switch which lights when dangerous reversed wiring condition exists. Simply flip switch to extinguish light and correct condition. Easy-access 10 amp fuse type 3 AG (rear mounted). Line voltage pilot light. Standard 1.8 meter heavy duty neoprene three-wire power cord (grounded to case) with NEMA plug. Input 120 volts, 60 cycles.
Weight: 794 g (1.75 lbs.)
Caution: As this is a solid state transformerless line voltage controller, it is NOT recommended for heaters rated less than 120 V.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty