Would you like to log in again?
CyroSaver Glycerol 30%, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CG89 CG89
89407-084PK 512.85 USD InStock
89407-084 CA89407-084
CyroSaver Glycerol 30%, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Culgenex™, CryoSaver™ with 30% glycerol is recommended for the preservation of laboratory microorganisms by freezing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty