Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, Ultrapure for molecular biology

Supplier: HARDY DIAGNOSTICS
Synonyms: Dihydrogen oxide

CG490BX CG490BX
89407-034PK 295.26 USD InStock
CA89407-034 89407-034
Water, Ultrapure for molecular biology
Water
CulGenex™ Ultra Pure, Molecular Grade Water is suitable for use in molecular biology applications that require water free of DNAses, RNAses, and proteases.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty