Would you like to log in again?
CRITERION™ Agar, Bacteriological Grade, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C5001 C5003 C5001 C5002 C5002 C5003
89405-070EA 4144.08 USD InStock
89405-070 CA89405-066 CA89405-068 89405-068 89405-066 CA89405-070
CRITERION™ Agar, Bacteriological Grade, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Agar, Bacteriological Grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.
  • Wide mouth opening reduces dust and allows easy scooping
  • Convenient handgrip with 'no slip' finger holds
  • Stackable containers
  • High solubility
  • Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty