Would you like to log in again?
SF Broth, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6933 C6932 C6930 C6931 C6931
89406-036EA 4167.28 USD InStock
89406-036 89406-034 CA89406-032 CA89406-030 89406-032
SF Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ SF Broth for the selective cultivation of fecal streptococci from milk, water, sewage and feces. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ SF Broth for the selective cultivation of fecal streptococci from milk, water, sewage and feces
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty