Would you like to log in again?
BBE Agar (Bacteroides Bile Esculin), Hardy Diagnostics
Supplier: Hardy Diagnostics
G05 G05
89407-132PK 97.82 USD InStock
89407-132 CA89407-132
BBE Agar (Bacteroides Bile Esculin), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and detection of Bacteroides fragilis group
BBE Agar, Bacteroides Bile Esculin, reducible, for anaerobes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty