Would you like to log in again?
Campy Cefe x Agar, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
A122 A122
89405-026PK 48.74 USD InStock
89405-026 CA89405-026
Campy Cefe x Agar, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of <italic>Campylobacter</italic> species such as<italic> C
jejuni</italic>, <italic>C. col</italic>, and <italic>C. lari</italic>. Contains lysed horse blood, cefoperazone and amphotercin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty