Would you like to log in again?
Lactose Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K137 U105 K137 K223 K223
89407-632PK 148 USD InStock
CA89407-632 CA89407-656 CA89407-640 89407-632 89407-640
Lactose Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of coliforms and Salmonella species in water, food, dairy, and pharmaceutical products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty