Would you like to log in again?
Malonate Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
K36 K36
89407-706PK 171.44 USD InStock
CA89407-706 89407-706
Malonate Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of gram negative bacilli based on malonate utilization.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty