Would you like to log in again?
Nitrate Reagent B, Naphthylamine, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z72 Z72
89407-772EA 28.88 USD InStock
CA89407-772 89407-772
Nitrate Reagent B, Naphthylamine, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For use in determining the nitrate reduction reaction of bacteria, other than Mycobacterium species.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty