Would you like to log in again?
Nitrate Reagent C, Zinc Dust, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z73
89407-774EA 30.06 USD InStock
89407-774
Nitrate Reagent C, Zinc Dust, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the completion of the nitrate reduction test
Confirms negatives after adding Nitrate A, product number Z124 or 89407-776 and Nitrate B, product number Z125 or 89407-772 reagents. True negatives will turn red after the zinc dust is added.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty