Would you like to log in again?
OxiSticks™ Oxidase Swabs, Swabs Per Vial, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z193 Z193
89407-794EA 34.26 USD InStock
CA89407-794 89407-794
OxiSticks™ Oxidase Swabs, Swabs Per Vial, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Rapid test for oxidase in bacteria
Simply touch the OxiStick™ Swab to the colony to be tested and observe for color change.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty