Would you like to log in again?
Orange Serum Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G181 G91 G91
89407-792PK 78.12 USD InStock
89407-792 89407-200 CA89407-792
Orange Serum Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and enumeration of aciduric bacteria and fungi by membrane filtration methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty