Would you like to log in again?
Phosphate Buffered Saline (PBS), Hardy Diagnostics
Supplier: Hardy Diagnostics
K146 K146
89407-798PK 238.89 USD InStock
CA89407-798 89407-798
Phosphate Buffered Saline (PBS), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Phosphate Buffered Saline (pH of 7.5) is recommended as a diluent for quantitative culturing or as a rinsing agent in microbiological, immunological, histological and molecular testing procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty