Would you like to log in again?
R2A Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G03 G03
89407-128PK 64.4 USD InStock
89407-128 CA89407-128
R2A Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
R2A Agar is recommended for use in the pour plate and spread plate methods for enumeration of heterotrophic bacteria in water, especially potable water.
Ordering information: A minimum order of 10 packs is required for purchase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty