Would you like to log in again?
Tryptone Glucose Extract Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G115 G115
89407-142PK 78.12 USD InStock
89407-142 CA89407-142
Tryptone Glucose Extract Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and enumeration of microorganisms found in food, water, and dairy products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty