Would you like to log in again?
Tryptic Soy Agar (TSA) with Lecithin and Tween, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C7132 C7131 C7133 C7131 C7132 C7133 C7130 C7130
89406-170EA 137.06 USD InStock
89406-170 CA89406-168 89406-168 CA89406-174 CA89406-172 89406-174 CA89406-170 89406-172
Tryptic Soy Agar (TSA) with Lecithin and Tween, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Tryptic Soy Agar with Lecithin and Tween® 80 is recommended for use in the detection and enumeration of microorganisms in replicate plating techniques
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Also used for the detection and enumeration of microorganisms present on the surfaces of sanitary importance. CRITERION™ Dehydrated Culture Media and container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty