Would you like to log in again?
Brucella Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
R18 R18
89407-404PK 111.95 USD InStock
89407-404 CA89407-404
Brucella Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Broth is recommended for use as a general purpose growth media for the isolation, cultivation, and differentiation of a variety of fastidious organisms, such as streptococcus and brucella species in clinical and food samples
Tube is optically clear, shatter resistant, polycarbonate tube.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty