Would you like to log in again?
Buffered Listeria Enrichment Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K183 K183 U233 U233
89407-422PK 205.02 USD InStock
89407-422 CA89407-422 CA89407-394 89407-394
Buffered Listeria Enrichment Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Buffered Listeria Enrichment Broth Base is used in procedures for the isolation of Listeria monocytogenes food samples
Listeria Enrichment Broth contains peptones and dextrose as a source of growth factors.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty