Would you like to log in again?
Campy CVA Agar with 5% Sheep Blood, Hardy Diagnostics
Supplier: Hardy Diagnostics
A40 A40
89405-034PK 41.53 USD InStock
CA89405-034 89405-034
Campy CVA Agar with 5% Sheep Blood, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of cephalothin resistant <italic>Campylobacter</italic> species such as<italic> C
jejuni</italic>, <italic>C. col</italic>, and <italic>C. lari</italic>. Contains lysed horse blood, cefoperazone and amphotercin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty