Would you like to log in again?
HardyCHROM™ O157, Hardy Diagnostics
Supplier: Hardy Diagnostics
G305 G305
89407-256PK 66.76 USD InStock
89407-256 CA89407-256
HardyCHROM™ O157, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A selective and differential medium recommended for the isolation of enterohemorrhagic E.coli O157 from food and environmental sources
Chromogenic substances in the media facilitate detection by colony color. Not for human diagnostic use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty