Would you like to log in again?
HardyCHROM™ Sakazakii, for Cronobacter sakazakii, Hardy Diagnostics
Supplier: Hardy Diagnostics
G315 G315
89407-262PK 117.21 USD InStock
CA89407-262 89407-262
HardyCHROM™ Sakazakii, for Cronobacter sakazakii, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
HardyCHROM™ sakazakii is a chromogenic medium recommended for the selective isolation and differention of Cronobacter (Enterobacter) sakazakii from other members of the family Enterobacteriaceae based on colony color
Especially useful for the screening infant formulas.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty