Would you like to log in again?
HardyCHROM™ Listeria, Hardy Diagnostics
Supplier: Hardy Diagnostics
G317 G317
89407-264PK 68.41 USD InStock
89407-264 CA89407-264
HardyCHROM™ Listeria, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
HardyCHROM™ Listeria is a chromogenic medium recommended for the selective isolation, differentiation, and enumeration of Listeria monocytogenes from food and environmental samples by colony color and appearance.
  • Modified and improved ALOA formula
  • Differentiates L. monocytogenes and L. ivanovii from other Listeria species
  • Colonies turn turquoise with a white halo for easy read-out
  • Results as early as 24 hours
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty