Would you like to log in again?
Iodine-Iodide Solution, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z129
89407-624EA 192.12 USD InStock
89407-624
Iodine-Iodide Solution, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Iodine-Iodide Solution to be added to Tetrathionate Broth, 89407-968 (sold separately)
Iodine-Iodide Solution, for Tetrathionate Broth.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty