Would you like to log in again?
Letheen Agar, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
G221 G221
89407-228PK 136.91 USD InStock
89407-228 CA89407-228
Letheen Agar, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Modified Letheen agar is used for evaluating the bactericidal activity, or microbial cleanliness, of a finished product.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty