Would you like to log in again?
PDX-SIB, Environmental Screen, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
2600325 2600325
89404-954PK 345.35 USD InStock
89404-954 CA89404-954
PDX-SIB, Environmental Screen, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Media for the detection of <italic>Salmonella</italic>, AOAC RI approved, 24-48 hour result, closed system, no special equipment needed, use with EnviroMAX sampling sponge on a stick 89405-022.
  • AOAC-RI approved
  • Closed system, once the broth media is inoculated the system stays closed
  • Three simple steps.
The easiest environmental <italic>Salmonella</italic> test available. This test is both reliable and inexpensive with presumptive results available for the most common <italic>Salmonella</italic> spp, within 24-48 hours, no sub-culturing or specimen transfers required.
Just collect your sample, inoculate the PDX-SIB media, and incubate. Sampling devices are not included.
<italic>Salmonella</italic> selective enrichment media containing a <italic>Salmonella</italic> -specific metabolic indicator.
Broth turns yellow when <italic>Salmonella</italic> species is present.
Excellent critical control point surveillance tool.
PDX-SIB was subjected to a panel of more than 100 <italic>Salmonella</italic> and non-<italic>Salmonell</italic>a</italic> organisms. The test exhibited 98.5% sensitivity with 95% specificity. The tests were conducted with inocula at both low (<10 CFU per sample) as well as high (>100 CFU per sample).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty