Would you like to log in again?
PIG® Gray Ham-O® Absorbent Mat Pad, New Pig
Supplier: New Pig
MAT117
56620-564PK 260.51 USD InStock
56620-564
PIG® Gray Ham-O® Absorbent Mat Pad, New Pig
Spill Control Materials
Exclusive "hamouflage" top layer saves on product and disposal costs; absorbed grime is invisible, so you'll leave mat in place longer and save money
Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated. Under the Ham-O top layer is the proven eight-layer, 100% polypropylene construction of PIG Mat. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Absorbs and retains oils, coolants, solvents and water. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Fast-wicking "diaper action" transfers liquids from the top layer into the absorbent inner layers; spills disappear in seconds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty