Would you like to log in again?
Pig Blue® Barrel Top Absorbent Mat, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
BLU255
89132-780CS 110.86 USD InStock
89132-780
Pig Blue® Barrel Top Absorbent Mat, New Pig
Spill Control Materials
For your biggest pump and dispensing messes, put the power of highly-absorbent Pig Blue Mat to work
Contains 75% recycled fibers; Excellent in situations where green products are desired or required. Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08.
Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Fits 55-gallon steel drum tops; pre-cut with holes for 3/4" and 2" bungs to provide a perfect fit. Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture.
Soak up pump drips without cluttering up your work space with dirty rags. Consistent construction produces a mat that is free of light spots to absorb liquid evenly for better wicking and retention. FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty